Folinic acid calcium salt, hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C2235-1G C2235-5G
TCC2235-1GEA 163.93 CAD
TCC2235-1G TCC2235-5G
Folinic acid calcium salt, hydrate ≥98.0% (by HPLC, titration analysis)
Folinic acid calcium salt

CAS Number: 1492-18-8 MDL Number: MFCD00006704 Molecular Formula: C20H23N7O7 Molecular Weight: 511.51 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal


Form: Crystal

Formula: C₂₀H₂₁CaN₇O₇
MDL Number: MFCD00006704
CAS Number: 1492-18-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=784da6b2c15b6004f4a2fe4dc87f3bd4cedf12cb&vid=6522c6d99adccea8a58203c6dffc97a39683a4c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR