trans-1,4-Cyclohexanedimethanol ≥98.0%

Supplier: TCI America

Synonyms: trans-1,4-Bis(hydroxymethyl)cyclohexane

C2234-5G C2234-25G
TCC2234-5GEA 94.8 CAD
TCC2234-5G TCC2234-25G
trans-1,4-Cyclohexanedimethanol ≥98.0%
trans-1,4-Cyclohexanedimethanol

CAS Number: 3236-48-4
MDL Number: MFCD00066360
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 164
Melting point (°C): 67
Flash Point (°C): 167


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: trans-1,4-Bis(hydroxymethyl)cyclohexane

Formula: C₈H₁₆O₂
Boiling Pt: 164 °C
Melting Pt: 67 °C
Flash Pt: 167 °C
MDL Number: MFCD00066360
CAS Number: 3236-48-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b104beea956d8b0749090f58e70d74e8627b1329&vid=b104beea956d8b0749090f58e70d74e8627b1329&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR