2,2'-O-Cyclouridine ≥98.0%
Supplier: TCI America
C2208-25G
C2208-5G
TCC2208-25GEA
204.93
CAD
TCC2208-25G
TCC2208-5G
2,2'-O-Cyclouridine ≥98.0%
2,2'-O-Cyclouridine
CAS Number: 3736-77-4
MDL Number: MFCD00004945
Molecular Formula: C9H10N2O5
Molecular Weight: 226.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 244
Specific rotation [a]20/D: -20 deg (C=4, H2O)
Storage Temperature: <0°C
Product Categories: Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: 2,2'-O-Anhydro-(1-beta-D-arabinofuranosyl)uracil
Formula:
C₉H₁₀N₂O₅ Melting Pt: 244 °C |
MDL Number:
MFCD00004945 CAS Number: 3736-77-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...