2,2'-O-Cyclocytidine hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
Ancitabine Hydrochloride, Cyclocytidine hydrochloride, (2R,3R,3aS,9aR)-2-(Hydroxymethyl)-6-imino-3,3a,6,9a-tetrahydro-2H-furo[2',3':4,5]oxazolo[3,2-a]pyrimidin-3-ol hydrochloride, 2,2'-Anhydro-(1-beta-D-arabinofuranosyl)cytosine Hydrochloride
CAS Number: 10212-25-6
MDL Number: MFCD00012636
Molecular Formula: C9H11N3O4
Molecular Weight: 261.66
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 264
Specific rotation [a]20/D: -21 deg (C=2, H2O)
Product Categories: Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: 2,2'-Anhydro-(1-beta-D-arabinofuranosyl)cytosine Hydrochloride, Ancitabine Hydrochloride
Formula:
C₉H₁₁N₃O₄ MW: 261.66 g/mol Melting Pt: 264 °C |
MDL Number:
MFCD00012636 CAS Number: 10212-25-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...