3-Aminopyrazine-2-carboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: 3-Amino-2-pyrazinecarboxylic acid

A0848-25G A0848-5G
TCA0848-25GEA 292.3 CAD
TCA0848-25G TCA0848-005G
3-Aminopyrazine-2-carboxylic acid ≥98.0%
3-Aminopyrazine-2-carboxylic acid

CAS Number: 5424-01-1
MDL Number: MFCD00006141
Molecular Formula: C5H5N3O2
Molecular Weight: 139.11
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 195

Formula: C₅H₅N₃O₂
Melting Pt: 195 °C
MDL Number: MFCD00006141
CAS Number: 5424-01-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d66ba5759e2401f3d20bbfd38df026d7732b8ac4&vid=d66ba5759e2401f3d20bbfd38df026d7732b8ac4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR