(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol ≥97.0%

Supplier: TCI America

Synonyms: 6-Chloropurine riboside

C2206-1G C2206-5G
TCC2206-1GEA 147.86 CAD
TCC2206-1G TCC2206-5G
(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol ≥97.0%
(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

CAS Number: 5399-87-1
MDL Number: MFCD00005738
Molecular Formula: C10H11ClN4O4
Molecular Weight: 286.67
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -45 deg (C=0.8, H2O)


Product Categories: Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: 6-Chloro-9-(beta-D-ribofuranosyl)purine

Formula: C₁₀H₁₁ClN₄O₄
MW: 286.67 g/mol
MDL Number: MFCD00005738
CAS Number: 5399-87-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6262f2368b15e8dd065ab176dc10f10d502f5620&vid=6262f2368b15e8dd065ab176dc10f10d502f5620&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR