Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II)
Supplier: TCI America
|
Warning
|
Synonyms:
Chlorocyclopentadienylbis(triphenylphosphine)ruthenium(II), Cyclopentadienylbis(triphenylphosphine)ruthenium(II) chloride, RuClCp(PPh3)
C2201-5G
C2201-1G
TCC2201-5GEA
358.66
CAD
TCC2201-5G
TCC2201-1G
Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II)
Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II)
CAS Number: 32993-05-8
MDL Number: MFCD00075004
Molecular Formula: C41H35ClP2Ru
Molecular Weight: 726.20
Form: Crystal
Color: Yellow
Melting point (°C): 135
Other Product Names / Synonyms: Chlorocyclopentadienylbis(triphenylphosphine)ruthenium(II)
Formula:
C₄₁H₃₅ClP₂Ru MW: 726.20 g/mol |
MDL Number:
MFCD00075004 CAS Number: 32993-05-8 |
|
Specification Test Results
Elemental analysis (Carbon) | 66.00 to 69.00 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...