1,2,4,5-Cyclohexanetetracarboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: Cyclohexane-1,2,4,5-tetracarboxylic acid

C2198-25G C2198-5G
TCC2198-25GEA 788.42 CAD
TCC2198-25G TCC2198-5G
1,2,4,5-Cyclohexanetetracarboxylic acid ≥98.0%
1,2,4,5-Cyclohexanetetracarboxylic acid

CAS Number: 15383-49-0
MDL Number: MFCD00435556
Molecular Formula: C10H12O8
Molecular Weight: 260.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 271

Formula: C₁₀H₁₂O₈
MW: 260.20 g/mol
Melting Pt: 271 °C
MDL Number: MFCD00435556
CAS Number: 15383-49-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=feba2c17a996892e9e58576d5f161a865841cf51&vid=feba2c17a996892e9e58576d5f161a865841cf51&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR