2-Chloro-5-iodobenzotrifluoride ≥98.0%

Supplier: TCI America

C2196-25G C2196-5G
TCC2196-25GEA 156.42 CAD
TCC2196-25G TCC2196-5G
2-Chloro-5-iodobenzotrifluoride ≥98.0%
2-Chloro-5-iodobenzotrifluoride

CAS Number: 260355-20-2
Molecular Formula: C7H3ClF3I
Molecular Weight: 306.45
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Freezing point (°C): 28


Other Product Names / Synonyms: 1-Chloro-4-iodo-2-(trifluoromethyl)benzene

Formula: C₇H₃ClF₃I
MDL Number: MFCD09037801
CAS Number: 260355-20-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48202c21b8df62aee4f27e95e6ad4d2b3c62410d&vid=48202c21b8df62aee4f27e95e6ad4d2b3c62410d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR