2-Chloroadenosine ≥98.0% for synthesis

Supplier: TCI America

Synonyms: (2R,3R,4S,5R)-2-(6-Amino-2-chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

C2192-100MG C2192-1G
TCC2192-100MGEA 42.66 CAD
TCC2192-100MG TCC2192-1G
2-Chloroadenosine ≥98.0% for synthesis
2-Chloroadenosine

CAS Number: 146-77-0
MDL Number: MFCD00005734
Molecular Formula: C10H12ClN5O4
Molecular Weight: 301.69
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 162
Specific rotation [a]20/D: -50 deg (C=0.1, H2O)


Product Categories: Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: 2-CADO, 6-Amino-2-chloropurine Riboside, 6-Amino-2-chloro-9-(beta-D-ribofuranosyl)purine

Formula: C₁₀H₁₂ClN₅O₄
MW: 301.69 g/mol
Melting Pt: 162 °C
MDL Number: MFCD00005734
CAS Number: 146-77-0
EINECS: 205-678-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b338c50fdb5b0b85be35920d57020c4493592d17&vid=b338c50fdb5b0b85be35920d57020c4493592d17&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR