3-Chloro-4-morpholino-1,2,5-thiadiazole ≥98.0%

Supplier: TCI America

Synonyms: Timolol Related Compound F, 4-(4-Chloro-1,2,5-thiadiazol-3-yl)morpholine

C2189-25G C2189-5G
TCC2189-25GEA 112.34 CAD
TCC2189-25G TCC2189-5G
3-Chloro-4-morpholino-1,2,5-thiadiazole ≥98.0%
3-Chloro-4-morpholino-1,2,5-thiadiazole

CAS Number: 30165-96-9
MDL Number: MFCD00274160
Molecular Formula: C6H8ClN3OS
Molecular Weight: 205.66
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 46
Flash Point (°C): 110

Formula: C₆H₈ClN₃OS
Melting Pt: 46 °C
Flash Pt: 110 °C
MDL Number: MFCD00274160
CAS Number: 30165-96-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42385a5781b0d9fac7784162f9e7070fb042d6ac&vid=42385a5781b0d9fac7784162f9e7070fb042d6ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR