Cyclohexylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
C2188-1G
C2188-5G
TCC2188-1GEA
40.21
CAD
TCC2188-1G
TCC2188-5G
Cyclohexylboronic acid (contains varying amounts of Anhydride)
Cyclohexylboronic acid
CAS Number: 4441-56-9
MDL Number: MFCD01311824
Molecular Formula: C6H13BO2
Molecular Weight: 127.98
Form: Crystal
Color: White
Melting point (°C): 120
Formula:
C₆H₁₃BO₂ Melting Pt: 120 °C |
MDL Number:
MFCD01311824 CAS Number: 4441-56-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...