1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) ≥98.0%

Supplier: TCI America

Synonyms: Cyclohexane-1,3-dicarboxylic acid, 1,3-Cyclohexanedicarboxylic acid

C2186-5G C2186-25G
TCC2186-5GEA 188.49 CAD
TCC2186-5G TCC2186-25G
1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) ≥98.0%
1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers)

CAS Number: 3971-31-1
MDL Number: MFCD00134411
Molecular Formula: C8H12O4
Molecular Weight: 172.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 149

Formula: C₈H₁₂O₄
MW: 172.18
Melting Pt: 149 °C
MDL Number: MFCD00134411
CAS Number: 3971-31-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=525589925dcb4a768e455f58fb9f5cf8a6d1c1fa&vid=231189542e4af61df5f0f6f96c14a55110fddea5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR