N(α)-Benzyloxycarbonyl-D-lysine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Cbz-D-Lys-OH, Z-D-Lys-OH

C2136-1G C2136-5G
TCC2136-1GEA 72.68 CAD
TCC2136-1G TCC2136-5G
N(α)-Benzyloxycarbonyl-D-lysine ≥98.0% (by HPLC, titration analysis)
N(α)-Benzyloxycarbonyl-D-lysine

CAS Number: 70671-54-4
MDL Number: MFCD00067713
Molecular Formula: C14H20N2O4
Molecular Weight: 280.32
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 218
Specific rotation [a]20/D: 13 deg (C=2, 0.2mol/L HCl)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Cbz-D-lysine, Z-D-Lys-OH

Formula: C₁₄H₂₀N₂O₄
MW: 280.3 g/mol
Melting Pt: 218 °C
MDL Number: MFCD00067713
CAS Number: 70671-54-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=509b0b844fb104a8e46b0faaa2676bce1bbbfd36&vid=509b0b844fb104a8e46b0faaa2676bce1bbbfd36&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR