N-Carbobenzoxy-D-leucine dicyclohexylammonium salt ≥98.0%

Supplier: TCI America

Synonyms: N-Cbz-D-leucine Dicyclohexylammonium Salt, Z-D-Leu-OH.DCHA

C2135-1G C2135-5G
TCC2135-1GEA 47.4 CAD
TCC2135-1G TCC2135-5G
N-Carbobenzoxy-D-leucine dicyclohexylammonium salt ≥98.0%
N-Carbobenzoxy-D-leucine dicyclohexylammonium salt

CAS Number: 7662-58-0
Molecular Formula: C14H19NO4
Molecular Weight: 446.63
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 154
Specific rotation [a]20/D: 7.5 deg (C=3, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-D-leucine Dicyclohexylammonium Salt, Z-D-Leu-OH.DCHA

Formula: C₁₄H₁₉NO₄
MW: 446.63 g/mol
Melting Pt: 154 °C
MDL Number: MFCD10566885
CAS Number: 7662-58-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e12b2d0f1aa358180423cbf319876ac9978398fe&vid=e12b2d0f1aa358180423cbf319876ac9978398fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR