N-Acetyl-L-tyrosine ethyl ester monohydrate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

A0118-1G A0118-5G
TCA0118-001GEA 31.6 CAD
TCA0118-001G TCA0118-005G
N-Acetyl-L-tyrosine ethyl ester monohydrate ≥98.0% (by HPLC, total nitrogen)
N-Acetyl-L-tyrosine ethyl ester monohydrate

CAS Number: 36546-50-6
MDL Number: MFCD00149093
Molecular Formula: C13H17NO4
Molecular Weight: 251.28
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 81
Specific rotation [a]20/D: 22 deg (C=1, EtOH)


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: Ac-Tyr-OEt

Formula: C₁₃H₁₇NO₄
MW: 269.297 g/mol
Melting Pt: 81 °C
MDL Number: MFCD00149093
CAS Number: 840-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e46352230004ee5f5db16fec99a3c615090d7631&vid=e46352230004ee5f5db16fec99a3c615090d7631&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR