2-Chloro-m-toluic acid ≥98.0%

Supplier: TCI America

C2114-25G C2114-5G
TCC2114-25GEA 218.04 CAD
TCC2114-25G TCC2114-5G
2-Chloro-m-toluic acid ≥98.0%
2-Chloro-m-toluic acid

CAS Number: 15068-35-6
MDL Number: MFCD00079736
Molecular Formula: C8H7ClO2
Molecular Weight: 170.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 143


Other Product Names / Synonyms: 2-Chloro-m-toluic Acid

Formula: C₈H₇ClO₂
MW: 170.6
Melting Pt: 143 °C
MDL Number: MFCD00079736
CAS Number: 15068-35-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8550891252df64fb0685684ff53342d01731318f&vid=8550891252df64fb0685684ff53342d01731318f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR