4'-Chloropivaloanilide ≥98.0%

Supplier: TCI America

Synonyms: N-(4-Chlorophenyl)pivalamide, 4-Chloro-N-pivaloylaniline, N-(4-Chlorophenyl)-2,2-dimethylpropionamide

C2099-250G C2099-25G
TCC2099-250GEA 515.71 CAD
TCC2099-250G TCC2099-25G
4'-Chloropivaloanilide ≥98.0%
4'-Chloropivaloanilide

CAS Number: 65854-91-3
MDL Number: MFCD01571825
Molecular Formula: C11H14ClNO
Molecular Weight: 211.69
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 152


Other Product Names / Synonyms: 4-Chloro-N-pivaloylaniline, N-(4-Chlorophenyl)pivalamide, N-(4-Chlorophenyl)-2,2-dimethylpropionamide

Formula: C₁₁H₁₄ClNO
MW: 211.69 g/mol
Melting Pt: 152 °C
MDL Number: MFCD01571825
CAS Number: 65854-91-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fde286fe108dadc24fd75a78bc678417672761d9&vid=fde286fe108dadc24fd75a78bc678417672761d9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR