1,1-Cyclohexanediacetic acid monoamide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C2096-25G C2096-5G
TCC2096-25GEA 310.3 CAD
TCC2096-25G TCC2096-5G
1,1-Cyclohexanediacetic acid monoamide ≥98.0% (by titrimetric analysis)
1,1-Cyclohexanediacetic acid monoamide

CAS Number: 99189-60-3
MDL Number: MFCD02181086
Molecular Formula: C10H17NO3
Molecular Weight: 199.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 147


Other Product Names / Synonyms: 1,1-Cyclohexanediacetic Acid Monoamide, 1-(2-Amino-2-oxoethyl)cyclohexaneacetic Acid

Formula: C₁₀H₁₇NO₃
Melting Pt: 147 °C
MDL Number: MFCD02181086
CAS Number: 99189-60-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9152b978003e8419cb1476862646d6777cdfdc1c&vid=9152b978003e8419cb1476862646d6777cdfdc1c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR