4-Acryloylmorpholine ≥98.0% stabilized

Supplier: TCI America

A0841-25ML A0841-500ML A0841-100ML
TCA0841-025MLEA 43.37 CAD
TCA0841-025ML TCA0841-500ML TCA0841-100ML
4-Acryloylmorpholine ≥98.0% stabilized
4-Acryloylmorpholine

(stabilized with MEHQ)
CAS Number: 5117-12-4
MDL Number: MFCD00047413
Molecular Formula: C7H11NO2
Molecular Weight: 141.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Flash Point (°C): 127
Specific Gravity (20/20): 1.12

Formula: C₇H₁₁NO₂
MW: 141.17 g/mol
Boiling Pt: 165 °C
Density: 1.12 (20 °C)
Flash Pt: 127 °C
MDL Number: MFCD00047413
CAS Number: 5117-12-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3328849e0a19b9601d3ac2d661d7d882cc424538&vid=3328849e0a19b9601d3ac2d661d7d882cc424538&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR