5-(Carbobenzoxyamino)valeric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C2067-25G C2067-5G
TCC2067-25GEA 1463.08 CAD
TCC2067-25G TCC2067-5G
5-(Carbobenzoxyamino)valeric acid ≥98.0% (by titrimetric analysis)
5-(Carbobenzoxyamino)valeric acid

CAS Number: 23135-50-4
Molecular Formula: C13H17NO4
Molecular Weight: 251.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 106


Other Product Names / Synonyms: 5-(Carbobenzoxyamino)pentanoic Acid, N-Cbz-5-aminovaleric Acid, N-Cbz-5-aminopentanoic Acid

Formula: C₁₃H₁₇NO₄
Melting Pt: 106 °C
MDL Number: MFCD00188458
CAS Number: 23135-50-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23ff39ac6f5b7f8abc246a6b006d7779a3f51bf5&vid=23ff39ac6f5b7f8abc246a6b006d7779a3f51bf5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR