(1ɑ,3ɑ,5ɑ)-1,3,5-Cyclohexanetricarboxylic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Cis,cis-cyclohexane-1,3,5-tricarboxylic acid, (1α,3α,5α)-1,3,5-Cyclohexanetricarboxylic acid
C2063-25G
C2063-5G
TCC2063-25GEA
752.7
CAD
TCC2063-25G
TCC2063-5G
(1ɑ,3ɑ,5ɑ)-1,3,5-Cyclohexanetricarboxylic acid ≥98.0% (by titrimetric analysis)
(1alpha,3alpha,5alpha)-1,3,5-Cyclohexanetricarboxylic acid
CAS Number: 16526-68-4
MDL Number: MFCD00075294
Molecular Formula: C9H12O6
Molecular Weight: 216.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 218
Formula:
C₉H₁₂O₆ MW: 216.19 g/mol Melting Pt: 218 °C |
MDL Number:
MFCD00075294 CAS Number: 16526-68-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...