trans-4-(Carbobenzoxyamino)cyclohexanecarboxylic acid ≥96.0%

Supplier: TCI America

Synonyms: trans-4-(Cbz-amino)cyclohexanecarboxylic Acid, trans-4-(((Benzyloxy)carbonyl)amino)cyclohexanecarboxylic acid, trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid

C2036-1G C2036-25G C2036-5G
TCC2036-1GEA 48.98 CAD
TCC2036-1G TCC2036-25G TCC2036-5G
trans-4-(Carbobenzoxyamino)cyclohexanecarboxylic acid ≥96.0%
trans-4-(Carbobenzoxyamino)cyclohexanecarboxylic acid

CAS Number: 34771-04-5
Molecular Formula: C15H19NO4
Molecular Weight: 277.32
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal
Melting point (°C): 217


Other Product Names / Synonyms: trans-4-(Cbz-amino)cyclohexanecarboxylic Acid

Formula: C₁₅H₁₉NO₄
MW: 277.32 g/mol
Melting Pt: 217 °C
MDL Number: MFCD01550737
CAS Number: 34771-04-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d626460377bfebdad28248ad55db58ff0d9144e9&vid=d626460377bfebdad28248ad55db58ff0d9144e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR