2-(4-Chlorophenyl)benzimidazole 98,0

Supplier: TCI America

Synonyms: 2-(4-Chlorophenyl)-1H-benzo[d]imidazole

C2027-1G C2027-5G
TCC2027-1GEA 229.1 CAD
TCC2027-1G TCC2027-5G
2-(4-Chlorophenyl)benzimidazole 98,0
2-(4-Chlorophenyl)benzimidazole

CAS Number: 1019-85-8
Molecular Formula: C13H9ClN2
Molecular Weight: 228.68
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 303

Formula: C₁₃H₉ClN₂
MW: 228.68 g/mol
Melting Pt: 303 °C
MDL Number: MFCD00224359
CAS Number: 1019-85-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba5927fbf0805985a3e17f1cc9eafe621c73d8f6&vid=ba5927fbf0805985a3e17f1cc9eafe621c73d8f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR