O-Acetyl-L-serine hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
(S)-3-Acetoxy-2-aminopropanoic acid hydrochloride, H-Ser(Ac)-OH.HCl
A0834-100MG
A0834-1G
TCA0834-100MGEA
66.04
CAD
TCA0834-100MG
TCA0834-1G
O-Acetyl-L-serine hydrochloride ≥98.0%
O-Acetyl-L-serine hydrochloride
CAS Number: 66638-22-0
MDL Number: MFCD00060169
Molecular Formula: C5H9NO4
Molecular Weight: 183.59
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 163
Specific rotation [a]20/D: 16 deg (C=5, EtOH)
Storage Temperature: 0-10°C
Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: H-Ser(Ac)-OH.HCl
Formula:
C₅H₉NO₄ MW: 183.59 g/mol Melting Pt: 163 °C |
MDL Number:
MFCD00060169 CAS Number: 66638-22-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...