Climbazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C2025-25G C2025-500G
TCC2025-25GEA 96.38 CAD
TCC2025-25G TCC2025-500G
Climbazole ≥98.0% (by HPLC, titration analysis)
Climbazole

CAS Number: 38083-17-9
Molecular Formula: C15H17ClN2O2
Molecular Weight: 292.76
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 98


Product Categories: Bioscience, Antifungal Ingredients
Other Product Names / Synonyms: 1-(4-Chlorophenoxy)-1-(imidazol-1-yl)-3,3-dimethyl-2-butanone

Formula: C₁₅H₁₇ClN₂O₂
Melting Pt: 98 °C
MDL Number: MFCD00055505
CAS Number: 38083-17-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da66b45f057f31571e11310d20d5c3dcc255cf24&vid=da66b45f057f31571e11310d20d5c3dcc255cf24&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR