Chloro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate ≥98.0% (by total nitrogen basis)
Supplier: TCI America
C1957-5G
C1957-1G
TCC1957-5GEA
81.48
CAD
TCC1957-5G
TCC1957-1G
Chloro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate ≥98.0% (by total nitrogen basis)
Chloro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate
CAS Number: 94790-35-9
Molecular Formula: C5H12ClF6N2P
Molecular Weight: 280.58
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 107
Storage Temperature: <0°C
Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification
Other Product Names / Synonyms: TCFH
Formula:
C₅H₁₂ClF₆N₂P Melting Pt: 107 °C |
MDL Number:
MFCD01862891 CAS Number: 94790-35-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...