(1S,2S)-(+)-1,2-Cyclohexanedicarboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: (1S,2S)-Cyclohexane-1,2-dicarboxylic acid

C1954-1G C1954-5G
TCC1954-1GEA 104.12 CAD
TCC1954-1G TCC1954-5G
(1S,2S)-(+)-1,2-Cyclohexanedicarboxylic acid ≥98.0%
(1S,2S)-(+)-1,2-Cyclohexanedicarboxylic acid

CAS Number: 21963-41-7
Molecular Formula: C8H12O4
Molecular Weight: 172.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 184
Specific rotation [a]20/D: 18 deg (C=1, Acetone)

Formula: C₈H₁₂O₄
Melting Pt: 184 °C
MDL Number: MFCD00151258
CAS Number: 21963-41-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=409e46274389695e93b6886c36c70f334c232ab6&vid=409e46274389695e93b6886c36c70f334c232ab6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR