(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (1R,2R)-Cyclohexane-1,2-dicarboxylic acid, (1R,2R)-(-)-trans-Cyclohexane-1,2-dicarboxylic acid

C1953-1G C1953-5G
TCC1953-1GEA 97.96 CAD
TCC1953-1G TCC1953-5G
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid ≥98.0% (by GC, titration analysis)
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid

CAS Number: 46022-05-3
Molecular Formula: C8H12O4
Molecular Weight: 172.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 184
Specific rotation [a]20/D: -18 deg (C=1, Acetone)

Formula: C₈H₁₂O₄
MW: 172.18
Melting Pt: 184 °C
MDL Number: MFCD00062973
CAS Number: 46022-05-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e0149b729a851f6abffd4b3b0a3a4f867504341&vid=2e0149b729a851f6abffd4b3b0a3a4f867504341&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR