3-(Carbobenzoxyamino)-1-propanol ≥98.0%

Supplier: TCI America

Synonyms: Benzyl N-(3-Hydroxypropyl)carbamate, N-(3-Hydroxypropyl)carbamic Acid Benzyl Ester, 3-(Cbz-amino)-1-propanol, Benzyl (3-hydroxypropyl)carbamate

C1932-25G C1932-5G
TCC1932-25GEA 205.4 CAD
TCC1932-25G TCC1932-5G
3-(Carbobenzoxyamino)-1-propanol ≥98.0%
3-(Carbobenzoxyamino)-1-propanol

CAS Number: 34637-22-4
MDL Number: MFCD01321351
Molecular Formula: C11H15NO3
Molecular Weight: 209.25
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 54
Flash Point (°C): 110


Other Product Names / Synonyms: Benzyl N-(3-Hydroxypropyl)carbamate, 3-(Cbz-amino)-1-propanol, N-(3-Hydroxypropyl)carbamic Acid Benzyl Ester

Formula: C₁₁H₁₅NO₃
Melting Pt: 54 °C
Flash Pt: 110 °C
MDL Number: MFCD01321351
CAS Number: 34637-22-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b30674e1b60e63ab10bbcff918b64793244c2609&vid=b30674e1b60e63ab10bbcff918b64793244c2609&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR