4-(Chloromethyl)-5-methyl-1,3-dioxol-2-one ≥94.0%
Supplier: TCI America
C1882-25G
C1882-5G
TCC1882-25GEA
464.52
CAD
TCC1882-25G
TCC1882-5G
4-(Chloromethyl)-5-methyl-1,3-dioxol-2-one ≥94.0%
4-(Chloromethyl)-5-methyl-1,3-dioxol-2-one
CAS Number: 80841-78-7
MDL Number: MFCD07787494
Molecular Formula: C5H5ClO3
Molecular Weight: 148.54
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 93
Flash Point (°C): 129
Specific Gravity (20/20): 1.36
Formula:
C₅H₅ClO₃ Boiling Pt: 93 °C Density: 1.36 (20 °C) Flash Pt: 129 °C |
MDL Number:
MFCD07787494 CAS Number: 80841-78-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...