2-Chloro-4'-fluorobenzophenone ≥98.0%

Supplier: TCI America

Synonyms: (2-Chlorophenyl)(4-fluorophenyl)methanone

C1860-25G C1860-5G
TCC1860-25GEA 365.3 CAD
TCC1860-25G TCC1860-5G
2-Chloro-4'-fluorobenzophenone ≥98.0%
2-Chloro-4'-fluorobenzophenone

CAS Number: 1806-23-1
MDL Number: MFCD00000559
Molecular Formula: C13H8ClFO
Molecular Weight: 234.65
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 62

Formula: C₁₃H₈ClFO
MW: 234.6534 g/mol
Melting Pt: 62 °C
MDL Number: MFCD00000559
CAS Number: 1806-23-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f78ee328be2036e18ed6371a8b93ed3f74beda9&vid=1f78ee328be2036e18ed6371a8b93ed3f74beda9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR