4-Chloro-1,3-dioxolan-2-one ≥95.0%

Supplier: TCI America

Synonyms: 4-Chloro-2-oxo-1,3-dioxolane, Chloroethylene Carbonate

C1858-25G C1858-5G
TCC1858-25GEA 620.94 CAD
TCC1858-25G TCC1858-5G
4-Chloro-1,3-dioxolan-2-one ≥95.0%
4-Chloro-1,3-dioxolan-2-one

CAS Number: 3967-54-2
MDL Number: MFCD00005383
Molecular Formula: C3H3ClO3
Molecular Weight: 122.50
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 123
Flash Point (°C): 113
Specific Gravity (20/20): 1.51
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: 4-Chloro-2-oxo-1,3-dioxolane, Chloroethylene Carbonate

Formula: C₃H₃ClO₃
MW: 122.50 g/mol
Boiling Pt: 123 °C
Density: 1.51 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00005383
CAS Number: 3967-54-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a73ee558f5e3f19061458400066c7a85c0c9fcff&vid=a73ee558f5e3f19061458400066c7a85c0c9fcff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR