3-Chloro-2,4(3H,5H)-furandione ≥98.0%

Supplier: TCI America

Synonyms: 3-Chlorotetronic Acid, 3-Chlorofuran-2,4(3H,5H)-dione, 3-Chlorotetrahydrofuran-2,4-dione, 3-Chloro-4-hydroxy-2(5H)-furanone

C1810-5G C1810-25G
TCC1810-5GEA 134.3 CAD
TCC1810-5G TCC1810-25G
3-Chloro-2,4(3H,5H)-furandione ≥98.0%
3-Chloro-2,4(3H,5H)-furandione

CAS Number: 4971-55-5
MDL Number: MFCD00134273
Molecular Formula: C4H3ClO3
Molecular Weight: 134.52
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal


Other Product Names / Synonyms: 3-Chloro-4-hydroxy-2(5H)-furanone, 3-Chlorotetrahydrofuran-2,4-dione, 3-Chlorotetronic Acid

Formula: C₄H₃ClO₃
MW: 134.52 g/mol
MDL Number: MFCD00134273
CAS Number: 4971-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10c0f6cec0c6cc8642e9a677dbff55978e31f9ac&vid=10c0f6cec0c6cc8642e9a677dbff55978e31f9ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR