1-(Carbamoylmethyl)pyridinium chloride ≥98.0%
Supplier: TCI America
Synonyms:
1-(2-Amino-2-oxoethyl)pyridin-1-ium chloride
A0829-25G
A0829-5G
TCA0829-025GEA
134.3
CAD
TCA0829-025G
TCA0829-005G
1-(Carbamoylmethyl)pyridinium chloride ≥98.0%
1-(Carbamoylmethyl)pyridinium chloride
CAS Number: 41220-29-5
MDL Number: MFCD00032002
Molecular Formula: C7H9ClN2O
Molecular Weight: 172.61
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 213
Formula:
C₇H₉ClN₂O Melting Pt: 213 °C |
MDL Number:
MFCD00032002 CAS Number: 41220-29-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...