N-Carbobenzoxy-2-nitrobenzenesulfonamide ≥98.0%

Supplier: TCI America

Synonyms: N-Cbz-2-nitrobenzenesulphonamide, N-Cbz-2-nitrobenzenesulfonamide

C1757-25G C1757-5G
TCC1757-25GEA 884.33 CAD
TCC1757-25G TCC1757-5G
N-Carbobenzoxy-2-nitrobenzenesulfonamide ≥98.0%
N-Carbobenzoxy-2-nitrobenzenesulfonamide

CAS Number: 245365-64-4
MDL Number: MFCD06797080
Molecular Formula: C14H12N2O6S
Molecular Weight: 336.32
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 145


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Amination, Bioscience, Chemical Building Blocks for Polyamines
Other Product Names / Synonyms: N-Cbz-2-nitrobenzenesulfonamide

Formula: C₁₄H₁₂N₂O₆S
MW: 336.32 g/mol
Melting Pt: 145 °C
MDL Number: MFCD06797080
CAS Number: 245365-64-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=655fe4fb50204cd87e18f893810da94c9daaef7d&vid=655fe4fb50204cd87e18f893810da94c9daaef7d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR