(2-Chlorophenyl)acetic acid methyl ester ≥98.0%

Supplier: TCI America

Synonyms: (2-Chlorophenyl)acetic acid methyll ester, Methyl 2-chlorophenylacetate, Methyl (2-Chlorophenyl)acetate, Methyl (2-chlorophenyl)acetate

C1740-25G C1740-100G
TCC1740-25GEA 93.22 CAD
TCC1740-25G TCC1740-100G
(2-Chlorophenyl)acetic acid methyl ester ≥98.0%
(2-Chlorophenyl)acetic acid methyl ester

CAS Number: 57486-68-7
MDL Number: MFCD00137445
Molecular Formula: C9H9ClO2
Molecular Weight: 184.62
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 128
Specific Gravity (20/20): 1.21


Other Product Names / Synonyms: (2-Chlorophenyl)acetic Acid Methyl Ester

Formula: (C₆H₄Cl)CH₂COOCH₃
MW: 184.62 g/mol
Boiling Pt: 128 °C
Density: 1.21 (20 °C)
MDL Number: MFCD00137445
CAS Number: 57486-68-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdacec3719222189457ec3ced096373566092707&vid=bdacec3719222189457ec3ced096373566092707&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR