4'-Chloro-2,2,2-trifluoroacetophenone ≥97.0%
Supplier: TCI America
C1738-1G
C1738-5G
TCC1738-1GEA
48.98
CAD
TCC1738-1G
TCC1738-5G
4'-Chloro-2,2,2-trifluoroacetophenone ≥97.0%
4'-Chloro-2,2,2-trifluoroacetophenone
CAS Number: 321-37-9
MDL Number: MFCD00018686
Molecular Formula: C8H4ClF3O
Molecular Weight: 208.56
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Flash Point (°C): 78
Freezing point (°C): 25
Formula:
C₈H₄ClF₃O Flash Pt: 78 °C |
MDL Number:
MFCD00018686 CAS Number: 321-37-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...