4'-Carboxybenzo-18-crown-6 (4'-carboxybenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C1714-100MG C1714-1G
TCC1714-100MGEA 300.2 CAD
TCC1714-100MG TCC1714-1G
4'-Carboxybenzo-18-crown-6 (4'-carboxybenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥97.0% (by GC, titration analysis)
4'-Carboxybenzo-18-crown-6 (4'-Carboxybenzo-1,4,7,10,13,16-hexoxacyclooctadecane)

CAS Number: 60835-75-8
Molecular Formula: C17H24O8
Molecular Weight: 356.37
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 125


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 2,3-(4-Carboxybenzo)-1,4,7,10,13,16-hexaoxacyclooctadec-2-ene

Formula: C₁₇H₂₄O₈
Melting Pt: 125 °C
MDL Number: MFCD00143277
CAS Number: 60835-75-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc52f9cde7ef69b1569cf2cc5d8fa4ce119462de&vid=fc52f9cde7ef69b1569cf2cc5d8fa4ce119462de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR