Diethyl-trans-4-cyclohexene-1,2-dicarboxylate ≥97.0%
Supplier: TCI America
Synonyms:
trans-1,2,3,6-Tetrahydrophthalic Acid Diethyl Ester, Diethyl trans-1,2,3,6-Tetrahydrophthalate, trans-4-Cyclohexene-1,2-dicarboxylic Acid Diethyl Ester
CAS Number: 5048-50-0
MDL Number: MFCD06799457
Molecular Formula: C12H18O4
Molecular Weight: 226.27
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Other Product Names / Synonyms: trans-4-Cyclohexene-1,2-dicarboxylic Acid Diethyl Ester, Diethyl trans-1,2,3,6-Tetrahydrophthalate, trans-1,2,3,6-Tetrahydrophthalic Acid Diethyl Ester
Formula:
C₁₂H₁₈O₄ |
MDL Number:
MFCD06799457 CAS Number: 5048-50-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...