2-Chloroquinoline-4-carboxylic acid ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

C1703-25G C1703-5G
TCC1703-25GEA 900.6 CAD
TCC1703-25G TCC1703-5G
2-Chloroquinoline-4-carboxylic acid ≥97.0% (by HPLC, titration analysis)
2-Chloroquinoline-4-carboxylic acid

CAS Number: 5467-57-2
MDL Number: MFCD00023940
Molecular Formula: C10H6ClNO2
Molecular Weight: 207.61
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal

Formula: C₁₀H₆ClNO₂
MW: 207.61 g/mol
MDL Number: MFCD00023940
CAS Number: 5467-57-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b983a982b6a2dcc0a6f8c8d63a19750ad200ad9b&vid=b983a982b6a2dcc0a6f8c8d63a19750ad200ad9b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR