4-Chloro-6-nitro-m-cresol ≥98.0%

Supplier: TCI America

C1684-25G C1684-5G
TCC1684-25GEA 1203.96 CAD
TCC1684-25G TCC1684-5G
4-Chloro-6-nitro-m-cresol ≥98.0%
4-Chloro-6-nitro-m-cresol

CAS Number: 7147-89-9
MDL Number: MFCD00007114
Molecular Formula: C7H6ClNO3
Molecular Weight: 187.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 134


Other Product Names / Synonyms: 4-Chloro-5-methyl-2-nitrophenol

Formula: C₇H₆ClNO₃
Melting Pt: 134 °C
MDL Number: MFCD00007114
CAS Number: 7147-89-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e854ab5706c81253fd8fe8c43da6664c44e0e763&vid=e854ab5706c81253fd8fe8c43da6664c44e0e763&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR