Cytidine-5'-monophosphate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 5'-Cytidylic Acid, 5'-CMP, ((2R,3S,4R,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate

C1675-5G C1675-1G
TCC1675-5GEA 262.94 CAD
TCC1675-5G TCC1675-1G
Cytidine-5'-monophosphate ≥98.0% (by HPLC)
Cytidine-5'-monophosphate

CAS Number: 63-37-6
MDL Number: MFCD00006544
Molecular Formula: C9H14N3O8P
Molecular Weight: 323.20
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 9.8 deg (C=1, 0.5mol/L Na2HPO4 sol.)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Nucleotides and Their Analogs
Other Product Names / Synonyms: 5'-Cytidylic Acid, 5'-CMP

Formula: C₉H₁₄N₃O₈P
MW: 323.20 g/mol
MDL Number: MFCD00006544
CAS Number: 63-37-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da9ef97850d86effd034326d49b2202314bbddc9&vid=16aa6809a45d7d58e713ccf89f1475afee00134e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR