(±)-trans-1,2-Cyclohexanedicarboxylic anhydride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
C1657-25G
C1657-5G
TCC1657-25GEA
229.1
CAD
TCC1657-25G
TCC1657-5G
(±)-trans-1,2-Cyclohexanedicarboxylic anhydride ≥98.0% (by GC, titration analysis)
(±)-trans-1,2-Cyclohexanedicarboxylic anhydride
CAS Number: 14166-21-3
Molecular Formula: C8H10O3
Molecular Weight: 154.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 146
Formula:
C₈H₁₀O₃ Melting Pt: 146 °C |
MDL Number:
MFCD00064345 CAS Number: 14166-21-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...