2-Chloro-1,3-dimethylimidazolinium hexafluorophosphate ≥98.0%
Supplier: TCI America
Synonyms:
CIP, 2-Chloro-1,3-dimethyl-4,5-dihydro-1H-imidazol-3-ium hexafluorophosphate(V)
C1651-25G
C1651-5G
TCC1651-25GEA
232.26
CAD
TCC1651-25G
TCC1651-005G
2-Chloro-1,3-dimethylimidazolinium hexafluorophosphate ≥98.0%
2-Chloro-1,3-dimethylimidazolinium hexafluorophosphate
CAS Number: 101385-69-7
MDL Number: MFCD00191914
Molecular Formula: C5H10ClF6N2P
Molecular Weight: 278.56
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 233
Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis
Other Product Names / Synonyms: CIP
Formula:
C₅H₁₀ClF₆N₂P MW: 278.56 g/mol Melting Pt: 233 °C |
MDL Number:
MFCD00191914 CAS Number: 101385-69-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...