Diethyl cyclohexane-1,1-dicarboxylate ≥95.0%

Supplier: TCI America

C1640-1G C1640-5G C1640-25G
TCC1640-001GEA 127.98 CAD
TCC1640-001G TCC1640-5G TCC1640-25G
Diethyl cyclohexane-1,1-dicarboxylate ≥95.0%
Diethyl cyclohexane-1,1-dicarboxylate

CAS Number: 1139-13-5
MDL Number: MFCD02093457
Molecular Formula: C12H20O4
Molecular Weight: 228.29
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: 1,1-Cyclohexanedicarboxylic Acid Diethyl Ester

Formula: C₁₂H₂₀O₄
Boiling Pt: 137 °C
Density: 1.04 (20 °C)
MDL Number: MFCD02093457
CAS Number: 1139-13-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0efde80e6b5d36d0d8cf47356a6533c899b2cac4&vid=0efde80e6b5d36d0d8cf47356a6533c899b2cac4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR