5'-Chloro-2'-hydroxy-4'-methylacetophenone ≥95.0%

Supplier: TCI America

C1629-5G C1629-25G
TCC1629-005GEA 31.6 CAD
TCC1629-005G TCC1629-025G
5'-Chloro-2'-hydroxy-4'-methylacetophenone ≥95.0%
5'-Chloro-2'-hydroxy-4'-methylacetophenone

CAS Number: 28480-70-8
MDL Number: MFCD00191920
Molecular Formula: C9H9ClO2
Molecular Weight: 184.62
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 137
Melting point (°C): 69


Other Product Names / Synonyms: 4-Acetyl-2-chloro-5-hydroxytoluene, 2-Acetyl-4-chloro-5-methylphenol

Formula: C₉H₉ClO₂
Boiling Pt: 137 °C
Melting Pt: 69 °C
MDL Number: MFCD00191920
CAS Number: 28480-70-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cb4f8116ffc2f9737854213c18c2155a4a97ac4&vid=7cb4f8116ffc2f9737854213c18c2155a4a97ac4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR