4-Chloro-o-xylene (mixture of 3 and 4 isomers) ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 4-Chloro-1,2-dimethylbenzene, 1-Chloro-3,4-dimethylbenzene

C1626-25G C1626-500G
TCC1626-025GEA 48.98 CAD
TCC1626-025G TCC1626-500G
4-Chloro-o-xylene (mixture of 3 and 4 isomers) ≥97.0% (by GC)
4-Chloro-o-xylene

MDL Number: MFCD00000596
Molecular Formula: C8H9Cl
Molecular Weight: 140.61
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: Chloro-1,2-dimethylbenzene

Formula: C₈H₉Cl
MW: 140.61 g/mol
Density: 1.07 (20 °C)
MDL Number: MFCD00000596
CAS Number: 615-60-1

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR