4-(Chloromethyl)thiazole hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
C1623-5G
C1623-25G
TCC1623-005GEA
90.85
CAD
TCC1623-005G
TCC1623-25G
4-(Chloromethyl)thiazole hydrochloride ≥98.0% (by HPLC, titration analysis)
4-(Chloromethyl)thiazole hydrochloride
CAS Number: 7709-58-2
MDL Number: MFCD02094738
Molecular Formula: C4H4ClNS
Molecular Weight: 170.05
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Formula:
C₄H₄ClNS |
MDL Number:
MFCD02094738 CAS Number: 7709-58-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...