1-(4-Chlorobenzhydryl)piperazine ≥98.0%
Supplier: TCI America
C1611-5G
C1611-1G
TCC1611-005GEA
216.46
CAD
TCC1611-005G
76219-676
1-(4-Chlorobenzhydryl)piperazine ≥98.0%
1-(4-Chlorobenzhydryl)piperazine
CAS Number: 303-26-4
MDL Number: MFCD00005965
Molecular Formula: C17H19ClN2
Molecular Weight: 286.80
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 75
Flash Point (°C): 110
Other Product Names / Synonyms: 1-(4-Chlorodiphenylmethyl)piperazine
Formula:
C₁₇H₁₉ClN₂ Boiling Pt: 180 °C Melting Pt: 75 °C Flash Pt: 110 °C |
MDL Number:
MFCD00005965 CAS Number: 303-26-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...